Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL16A1
Entrez Gene ID
1307
Gene Name
collagen type XVI alpha 1 chain
Gene Aliases
447AA, FP1572
Chromosome Location
Chr.1: 31652247 - 31704167 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001856.3 NP_001847.3 70 - 71 4984 81
XM_005270481.1 XP_005270538.1 69 - 70 4931 81
XM_011540722.1 XP_011539024.1 69 - 70 4934 81
XM_011540723.1 XP_011539025.1 68 - 69 4886 81
XM_011540724.1 XP_011539026.1 69 - 70 4856 81
XM_011540726.2 XP_011539028.1 65 - 66 4402 81
XM_011540727.2 XP_011539029.1 60 - 61 3740 81
XM_011540728.2 XP_011539030.1 48 - 49 3222 81
XM_017000338.1 XP_016855827.1 69 - 70 4946 81
XM_017000339.1 XP_016855828.1 69 - 70 4943 81
XM_017000340.1 XP_016855829.1 47 - 48 3185 81
XM_017000341.1 XP_016855830.1 47 - 48 2975 81
GenBank mRNA AB209571.1 - 64 - 65 4198 81
AF370368.1 - 14 - 15 2168 81
M92642.1 - 71 - 72 4769 81
S57132.1 - 53 - 54 3365 81

View Full Product Details