The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012316.4 | NP_036448.1 | 13 - 14 | 1515 | 111 |
XM_005270711.3 | XP_005270768.1 | 13 - 14 | 1876 | 111 | |
GenBank mRNA | AF060543.1 | - | 13 - 14 | 1482 | 111 |
AK002111.1 | - | 13 - 14 | 1502 | 111 | |
AK128609.1 | - | 13 - 14 | 1478 | 111 | |
AK295615.1 | - | 13 - 14 | 1630 | 111 | |
AK301714.1 | - | 13 - 14 | 1469 | 111 | |
AK315490.1 | - | 13 - 14 | 1502 | 111 | |
BC020520.1 | - | 13 - 14 | 1490 | 111 | |
BT009843.1 | - | 12 - 13 | 1427 | 111 |