The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204077.1 | NP_001191006.1 | 4 - 5 | 541 | 94 |
NM_004788.3 | NP_004779.2 | 4 - 5 | 541 | 94 | |
GenBank mRNA | AF087968.1 | - | 4 - 5 | 480 | 94 |
AK296369.1 | - | 4 - 5 | 488 | 94 | |
BC030973.1 | - | 4 - 5 | 490 | 94 | |
BC052643.1 | - | 4 - 5 | 499 | 94 | |
BC062695.1 | - | 4 - 5 | 521 | 94 | |
BC111417.1 | - | 4 - 5 | 515 | 94 | |
BC112367.1 | - | 4 - 5 | 518 | 94 | |
BU167020.1 | - | 4 - 5 | 498 | 94 | |
D50916.1 | - | 4 - 5 | 478 | 94 |