The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134438.1 | NP_001127910.1 | 1 - 2 | 395 | 139 |
NM_145753.2 | NP_665696.1 | 1 - 2 | 395 | 139 | |
GenBank mRNA | AK096151.1 | - | 1 - 2 | 337 | 139 |
AK315050.1 | - | 1 - 2 | 307 | 139 | |
BC069194.1 | - | 1 - 2 | 284 | 139 |