The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134438.1 | NP_001127910.1 | 5 - 6 | 2411 | 101 |
NM_001134439.1 | NP_001127911.1 | 5 - 6 | 2172 | 101 | |
GenBank mRNA | AB209903.1 | - | 5 - 6 | 2053 | 101 |
AJ496194.1 | - | 4 - 5 | 2026 | 101 | |
AK092996.1 | - | 5 - 6 | 2097 | 101 | |
AK125555.1 | - | 4 - 5 | 780 | 101 | |
AK315050.1 | - | 5 - 6 | 2322 | 101 | |
BC142678.1 | - | 4 - 5 | 2018 | 101 | |
BC150210.1 | - | 4 - 5 | 2018 | 101 |