Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMAA
Entrez Gene ID
166785
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblA type
Gene Aliases
cblA
Chromosome Location
Chr.4: 145619388 - 145660035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172250.2 NP_758454.1 5 - 6 904 96
XM_011531684.2 XP_011529986.1 5 - 6 982 96
XM_011531685.2 XP_011529987.1 5 - 6 1025 96
XM_011531686.2 XP_011529988.1 4 - 5 1172 96
GenBank mRNA AK126662.1 - 5 - 6 904 96
BC101178.2 - 4 - 5 866 96
BC101179.2 - 4 - 5 866 96
BC101181.2 - 4 - 5 866 96

View Full Product Details