Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAIR1
Entrez Gene ID
3903
Gene Name
leukocyte associated immunoglobulin like receptor 1
Gene Aliases
CD305, LAIR-1
Chromosome Location
Chr.19: 54353624 - 54370556 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289023.2 NP_001275952.2 6 - 7 883 72
NM_001289025.2 NP_001275954.2 7 - 8 934 72
NM_001289026.2 NP_001275955.2 7 - 8 885 72
NM_001289027.2 NP_001275956.2 6 - 7 852 72
NM_002287.5 NP_002278.2 7 - 8 937 72
NM_021706.4 NP_068352.2 6 - 7 886 72
NR_110279.2 7 - 8 843 72
NR_110280.2 7 - 8 1178 72
GenBank mRNA AF013249.1 - 7 - 8 695 72
AF109683.1 - 6 - 7 660 72
AF251509.2 - 6 - 7 576 72
AK093559.1 - 2 - 3 902 72
AK097869.1 - 6 - 7 818 72
AK290415.1 - 7 - 8 751 72
AK303261.1 - 7 - 8 781 72
AK315419.1 - 7 - 8 783 72
BC027899.1 - 7 - 8 687 72
BI756388.1 - 7 - 8 599 72
CR542051.1 - 7 - 8 627 72

View Full Product Details