The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317949.1 | NP_001304878.1 | 5 - 6 | 572 | 101 |
NM_144725.3 | NP_653326.3 | 5 - 6 | 637 | 101 | |
XM_011513984.2 | XP_011512286.1 | 5 - 6 | 574 | 101 | |
XM_011513985.2 | XP_011512287.1 | 5 - 6 | 574 | 101 | |
XM_011513987.2 | XP_011512289.1 | 5 - 6 | 574 | 101 | |
XM_017009119.1 | XP_016864608.1 | 5 - 6 | 818 | 101 | |
XM_017009120.1 | XP_016864609.1 | 5 - 6 | 637 | 101 | |
XM_017009121.1 | XP_016864610.1 | 5 - 6 | 682 | 101 | |
XM_017009122.1 | XP_016864611.1 | 5 - 6 | 753 | 101 | |
XM_017009123.1 | XP_016864612.1 | 5 - 6 | 574 | 101 | |
XM_017009124.1 | XP_016864613.1 | 3 - 4 | 514 | 101 | |
XM_017009125.1 | XP_016864614.1 | 3 - 4 | 315 | 101 | |
XM_017009126.1 | XP_016864615.1 | 3 - 4 | 448 | 101 | |
GenBank mRNA | AK058168.1 | - | 5 - 6 | 637 | 101 |
AK293829.1 | - | 3 - 4 | 333 | 101 | |
AY498865.1 | - | 4 - 5 | 534 | 101 | |
BC057775.1 | - | 5 - 6 | 609 | 101 | |
JN799997.1 | - | 5 - 6 | 572 | 101 |