Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ULK4
Entrez Gene ID
54986
Gene Name
unc-51 like kinase 4
Gene Aliases
FAM7C1, REC01035
Chromosome Location
Chr.3: 41246599 - 41963020 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322500.1 NP_001309429.1 32 - 33 3693 72
NM_001322501.1 NP_001309430.1 31 - 32 3617 72
NM_017886.3 NP_060356.2 32 - 33 3693 72
NR_136342.1 30 - 31 3528 72
XM_011533874.2 XP_011532176.1 32 - 33 4271 72
XM_011533880.2 XP_011532182.1 16 - 17 2288 72
XM_017006711.1 XP_016862200.1 31 - 32 4187 72
XM_017006712.1 XP_016862201.1 32 - 33 4268 72
GenBank mRNA AK093396.1 - 11 - 12 945 72
AK304341.1 - 32 - 33 3324 72
AL133104.1 - 4 - 5 365 72
BC040739.1 - 4 - 5 269 72

View Full Product Details