The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105513.2 | NP_001098983.2 | 15 - 16 | 2260 | 78 |
NM_145755.2 | NP_665698.2 | 16 - 17 | 2404 | 78 | |
XM_005264921.4 | XP_005264978.1 | 16 - 17 | 2333 | 78 | |
XM_005264922.4 | XP_005264979.1 | 16 - 17 | 2333 | 78 | |
XM_005264923.3 | XP_005264980.1 | 16 - 17 | 2333 | 78 | |
XM_005264925.4 | XP_005264982.1 | 15 - 16 | 2210 | 78 | |
XM_005264927.4 | XP_005264984.1 | 12 - 13 | 1787 | 78 | |
XM_006713011.3 | XP_006713074.1 | 16 - 17 | 2333 | 78 | |
XM_011533447.2 | XP_011531749.1 | 15 - 16 | 2234 | 78 | |
GenBank mRNA | AJ487015.1 | - | 16 - 17 | 2366 | 78 |
AK093313.1 | - | 6 - 7 | 944 | 78 | |
AK302408.1 | - | 15 - 16 | 2243 | 78 | |
AK302465.1 | - | 15 - 16 | 2114 | 78 | |
BC129948.1 | - | 16 - 17 | 2315 | 78 |