The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286367.1 | NP_001273296.1 | 4 - 5 | 1050 | 133 |
NM_174913.2 | NP_777573.1 | 4 - 5 | 1050 | 133 | |
XM_005267385.1 | XP_005267442.1 | 4 - 5 | 1077 | 133 | |
XM_011536526.1 | XP_011534828.1 | 4 - 5 | 869 | 133 | |
XM_011536527.2 | XP_011534829.1 | 4 - 5 | 1077 | 133 | |
GenBank mRNA | AB095941.1 | - | 4 - 5 | 1042 | 133 |
BC025332.1 | - | 4 - 5 | 1028 | 133 | |
BX248018.1 | - | 4 - 5 | 1012 | 133 | |
BX647311.1 | - | 4 - 5 | 1017 | 133 |