The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020346.2 | NP_065079.1 | 3 - 4 | 872 | 108 |
GenBank mRNA | AB032435.1 | - | 3 - 4 | 872 | 108 |
AK294502.1 | - | 2 - 3 | 242 | 108 | |
BC069629.1 | - | 3 - 4 | 618 | 108 | |
BC069640.1 | - | 3 - 4 | 619 | 108 | |
BC069646.1 | - | 3 - 4 | 619 | 108 |