Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECEL1
Entrez Gene ID
9427
Gene Name
endothelin converting enzyme like 1
Gene Aliases
DA5D, DINE, ECEX, XCE
Chromosome Location
Chr.2: 232479827 - 232487859 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290787.1 NP_001277716.1 16 - 17 2388 59
NM_004826.3 NP_004817.2 16 - 17 2394 59
GenBank mRNA AB030579.1 - 15 - 16 2464 59
AB073895.1 - 15 - 16 1259 59
AY358923.1 - 16 - 17 2357 59
BC050453.1 - 16 - 17 2363 59
DQ114476.1 - 15 - 16 2146 59
Y16187.1 - 16 - 17 2357 59

View Full Product Details