Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GIGYF2
Entrez Gene ID
26058
Gene Name
GRB10 interacting GYF protein 2
Gene Aliases
GYF2, PARK11, PERQ2, PERQ3, TNRC15
Chromosome Location
Chr.2: 232697305 - 232860577 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103146.1 NP_001096616.1 11 - 12 1295 110
NM_001103148.1 NP_001096618.1 10 - 11 1211 110
NM_015575.3 NP_056390.2 13 - 14 1435 110
NR_103492.1 10 - 11 1211 110
GenBank mRNA AK001739.1 - 13 - 14 1409 110
AK302998.1 - 5 - 6 797 110
AY176045.1 - 13 - 14 1409 110
BC043402.1 - 11 - 12 1230 110
BC136251.1 - 10 - 11 1166 110
BC143352.1 - 10 - 11 1166 110
BX537885.1 - 11 - 12 1230 110
BX538172.1 - 10 - 11 1179 110
BX538321.1 - 11 - 12 1251 110
BX647505.1 - 10 - 11 1042 110

View Full Product Details