Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIGYF2
Entrez Gene ID
26058
Gene Name
GRB10 interacting GYF protein 2
Gene Aliases
GYF2, PARK11, PERQ2, PERQ3, TNRC15
Chromosome Location
Chr.2: 232697305 - 232860577 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103146.1 NP_001096616.1 14 - 15 1834 87
NM_001103147.1 NP_001096617.1 16 - 17 1967 87
NM_001103148.1 NP_001096618.1 13 - 14 1750 87
NM_015575.3 NP_056390.2 16 - 17 1974 87
NR_103492.1 13 - 14 1750 87
GenBank mRNA AB014542.2 - 16 - 17 1926 87
AK001739.1 - 16 - 17 1948 87
AK299418.1 - 15 - 16 1878 87
AK302998.1 - 8 - 9 1336 87
AY176045.1 - 16 - 17 1948 87
BC014047.2 - 1 - 2 158 87
BC043402.1 - 14 - 15 1769 87
BC136251.1 - 13 - 14 1705 87
BC143352.1 - 13 - 14 1705 87
BC146775.1 - 16 - 17 1926 87
BX537885.1 - 14 - 15 1769 87
BX538172.1 - 13 - 14 1718 87
BX538321.1 - 14 - 15 1790 87
BX647505.1 - 13 - 14 1581 87

View Full Product Details