The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103146.1 | NP_001096616.1 | 26 - 27 | 3659 | 137 |
NM_001103147.1 | NP_001096617.1 | 28 - 29 | 3792 | 137 | |
NM_001103148.1 | NP_001096618.1 | 25 - 26 | 3575 | 137 | |
NM_015575.3 | NP_056390.2 | 28 - 29 | 3799 | 137 | |
GenBank mRNA | AB014542.2 | - | 28 - 29 | 3751 | 137 |
AK027021.1 | - | 5 - 6 | 944 | 137 | |
AY176045.1 | - | 28 - 29 | 3773 | 137 | |
BC008072.2 | - | 2 - 3 | 303 | 137 | |
BC136251.1 | - | 25 - 26 | 3530 | 137 | |
BC146775.1 | - | 28 - 29 | 3751 | 137 | |
BX537885.1 | - | 26 - 27 | 3594 | 137 | |
BX538172.1 | - | 25 - 26 | 3543 | 137 | |
BX647172.1 | - | 10 - 11 | 2497 | 137 |