The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103146.1 | NP_001096616.1 | 7 - 8 | 691 | 85 |
NM_001103148.1 | NP_001096618.1 | 6 - 7 | 625 | 85 | |
NM_015575.3 | NP_056390.2 | 9 - 10 | 831 | 85 | |
NR_103492.1 | 6 - 7 | 607 | 85 | ||
GenBank mRNA | AK001739.1 | - | 9 - 10 | 805 | 85 |
AK299418.1 | - | 8 - 9 | 735 | 85 | |
AY176045.1 | - | 9 - 10 | 805 | 85 | |
BC043402.1 | - | 7 - 8 | 644 | 85 | |
BC136251.1 | - | 6 - 7 | 562 | 85 | |
BC143352.1 | - | 6 - 7 | 562 | 85 | |
BX537885.1 | - | 7 - 8 | 626 | 85 | |
BX538172.1 | - | 6 - 7 | 593 | 85 | |
BX538321.1 | - | 7 - 8 | 647 | 85 | |
BX647505.1 | - | 6 - 7 | 438 | 85 |