Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCNB2
Entrez Gene ID
9133
Gene Name
cyclin B2
Gene Aliases
HsT17299
Chromosome Location
Chr.15: 59105085 - 59125045 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004701.3 NP_004692.1 2 - 3 340 64
GenBank mRNA AB020981.1 - 2 - 3 225 64
AF002822.1 - 2 - 3 175 64
AF087910.1 - 2 - 3 282 64
AK001404.1 - 2 - 3 304 64
AK222611.1 - 2 - 3 273 64
AL080146.1 - 2 - 3 293 64
AM851054.1 - 2 - 3 156 64
BC105086.1 - 2 - 3 288 64
BC105112.1 - 2 - 3 288 64
BT019458.1 - 2 - 3 156 64
BU934242.1 - 2 - 3 341 64
CR533527.1 - 2 - 3 156 64

View Full Product Details