The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004701.3 | NP_004692.1 | 5 - 6 | 784 | 84 |
GenBank mRNA | AB020981.1 | - | 5 - 6 | 669 | 84 |
AF002822.1 | - | 5 - 6 | 619 | 84 | |
AF087910.1 | - | 5 - 6 | 84 | ||
AK001404.1 | - | 5 - 6 | 748 | 84 | |
AK222611.1 | - | 5 - 6 | 717 | 84 | |
AL080146.1 | - | 5 - 6 | 737 | 84 | |
AM851054.1 | - | 5 - 6 | 600 | 84 | |
BC105086.1 | - | 5 - 6 | 732 | 84 | |
BC105112.1 | - | 5 - 6 | 732 | 84 | |
BT019458.1 | - | 5 - 6 | 600 | 84 | |
CR533527.1 | - | 5 - 6 | 600 | 84 |