The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004139.4 | NP_004130.2 | 12 - 13 | 1371 | 159 |
GenBank mRNA | AF105067.1 | - | 12 - 13 | 1282 | 159 |
AK313625.1 | - | 12 - 13 | 1301 | 159 | |
BC022256.1 | - | 12 - 13 | 1300 | 159 | |
BQ028088.1 | - | 3 - 4 | 159 | ||
M35533.1 | - | 12 - 13 | 1253 | 159 |