Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP24
Entrez Gene ID
10893
Gene Name
matrix metallopeptidase 24
Gene Aliases
MMP-24, MMP25, MT-MMP 5, MT-MMP5, MT5-MMP, MT5MMP, MTMMP5
Chromosome Location
Chr.20: 35226736 - 35277001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006690.3 NP_006681.1 5 - 6 977 85
XM_011528500.2 XP_011526802.1 5 - 6 974 85
XM_017027598.1 XP_016883087.1 5 - 6 948 85
GenBank mRNA AB021227.1 - 5 - 6 977 85
AF131284.1 - 5 - 6 974 85
BC047614.1 - 5 - 6 917 85
DQ300264.1 - 6 - 7 818 85

View Full Product Details