The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006690.3 | NP_006681.1 | 6 - 7 | 1197 | 69 |
XM_011528500.2 | XP_011526802.1 | 6 - 7 | 1194 | 69 | |
XM_017027597.1 | XP_016883086.1 | 5 - 6 | 1032 | 69 | |
XM_017027598.1 | XP_016883087.1 | 6 - 7 | 1168 | 69 | |
GenBank mRNA | AB021227.1 | - | 6 - 7 | 1197 | 69 |
AF131284.1 | - | 6 - 7 | 1194 | 69 | |
BC047614.1 | - | 6 - 7 | 1137 | 69 | |
DQ300264.1 | - | 7 - 8 | 1038 | 69 |