The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317122.1 | NP_001304051.1 | 16 - 17 | 2388 | 66 |
NM_001317123.1 | NP_001304052.1 | 16 - 17 | 2232 | 66 | |
NM_012199.4 | NP_036331.1 | 16 - 17 | 2388 | 66 | |
XM_011541236.2 | XP_011539538.1 | 16 - 17 | 2358 | 66 | |
XM_017001016.1 | XP_016856505.1 | 16 - 17 | 2358 | 66 | |
XM_017001017.1 | XP_016856506.1 | 16 - 17 | 2349 | 66 | |
XM_017001018.1 | XP_016856507.1 | 13 - 14 | 1833 | 66 | |
XM_017001019.1 | XP_016856508.1 | 16 - 17 | 2446 | 66 | |
GenBank mRNA | AF093097.1 | - | 16 - 17 | 2374 | 66 |
AK002003.1 | - | 2 - 3 | 210 | 66 | |
AK314148.1 | - | 16 - 17 | 2349 | 66 | |
BC063275.1 | - | 16 - 17 | 2368 | 66 |