The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317122.1 | NP_001304051.1 | 7 - 8 | 1099 | 54 |
NM_001317123.1 | NP_001304052.1 | 7 - 8 | 943 | 54 | |
NM_012199.4 | NP_036331.1 | 7 - 8 | 1099 | 54 | |
XM_011541236.2 | XP_011539538.1 | 7 - 8 | 1060 | 54 | |
XM_017001016.1 | XP_016856505.1 | 7 - 8 | 1060 | 54 | |
XM_017001017.1 | XP_016856506.1 | 7 - 8 | 1060 | 54 | |
XM_017001018.1 | XP_016856507.1 | 4 - 5 | 535 | 54 | |
XM_017001019.1 | XP_016856508.1 | 7 - 8 | 1157 | 54 | |
GenBank mRNA | AF093097.1 | - | 7 - 8 | 1085 | 54 |
AK001622.1 | - | 7 - 8 | 943 | 54 | |
AK314148.1 | - | 7 - 8 | 1060 | 54 | |
BC063275.1 | - | 7 - 8 | 1079 | 54 |