The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014817.3 | NP_055632.2 | 1 - 1 | 3814 | 134 |
GenBank mRNA | AB014544.1 | - | 1 - 1 | 3693 | 134 |