The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021833.4 | NP_068605.1 | 1 - 2 | 199 | 53 |
XM_005263206.3 | XP_005263263.1 | 1 - 2 | 308 | 53 | |
XM_011532228.2 | XP_011530530.1 | 1 - 2 | 308 | 53 | |
GenBank mRNA | BC069556.1 | - | 1 - 2 | 199 | 53 |
BC096736.1 | - | 1 - 2 | 199 | 53 | |
BC098168.1 | - | 1 - 2 | 199 | 53 | |
BC098258.1 | - | 1 - 2 | 199 | 53 | |
BC098352.1 | - | 1 - 2 | 199 | 53 | |
U28480.1 | - | 1 - 2 | 123 | 53 |