The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021833.4 | NP_068605.1 | 4 - 5 | 708 | 138 |
XM_005263206.3 | XP_005263263.1 | 4 - 5 | 814 | 138 | |
XM_011532228.2 | XP_011530530.1 | 4 - 5 | 817 | 138 | |
GenBank mRNA | BC069556.1 | - | 4 - 5 | 708 | 138 |
BC096736.1 | - | 4 - 5 | 705 | 138 | |
BC098168.1 | - | 4 - 5 | 708 | 138 | |
BC098258.1 | - | 4 - 5 | 708 | 138 | |
BC098352.1 | - | 4 - 5 | 705 | 138 | |
U28480.1 | - | 4 - 5 | 632 | 138 |