The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152565.1 | NP_689778.1 | 3 - 4 | 549 | 92 |
GenBank mRNA | AK096027.1 | - | 3 - 4 | 549 | 92 |
AY079172.1 | - | 3 - 4 | 530 | 92 | |
BC065207.1 | - | 3 - 4 | 555 | 92 |