The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001211.5 | NP_001202.4 | 6 - 7 | 969 | 72 |
GenBank mRNA | AB208782.1 | - | 6 - 7 | 882 | 72 |
AF035933.1 | - | 6 - 7 | 72 | ||
AF046079.2 | - | 6 - 7 | 799 | 72 | |
AF046918.1 | - | 6 - 7 | 887 | 72 | |
AF053306.1 | - | 6 - 7 | 941 | 72 | |
AF068760.1 | - | 6 - 7 | 863 | 72 | |
AF107297.1 | - | 6 - 7 | 894 | 72 | |
AK296795.1 | - | 6 - 7 | 773 | 72 | |
AK296984.1 | - | 6 - 7 | 953 | 72 | |
AK312709.1 | - | 6 - 7 | 937 | 72 | |
BC018739.1 | - | 6 - 7 | 891 | 72 |