The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022776.4 | NP_073613.2 | 10 - 11 | 2576 | 80 |
GenBank mRNA | AF392454.1 | - | 10 - 11 | 2147 | 80 |
AK023074.1 | - | 5 - 6 | 973 | 80 | |
AK023226.1 | - | 5 - 6 | 1006 | 80 | |
AK292702.1 | - | 10 - 11 | 1928 | 80 | |
BC010422.1 | - | 5 - 6 | 1160 | 80 | |
BC065213.1 | - | 10 - 11 | 1926 | 80 |