The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042576.1 | NP_001036041.1 | 19 - 20 | 2632 | 101 |
NM_004587.2 | NP_004578.2 | 18 - 19 | 2555 | 101 | |
GenBank mRNA | AB037819.2 | - | - | 4311 | 101 |
AB287347.2 | - | 15 - 16 | 3967 | 101 | |
AF006751.1 | - | 18 - 19 | 2348 | 101 | |
AK093465.1 | - | 6 - 7 | 101 | ||
AL833822.1 | - | 16 - 17 | 2343 | 101 | |
BC000099.2 | - | 13 - 14 | 1313 | 101 | |
BC020482.1 | - | 15 - 16 | 1580 | 101 | |
BC080526.1 | - | 14 - 15 | 1439 | 101 | |
BC128577.1 | - | 18 - 19 | 2741 | 101 | |
BC128578.1 | - | 17 - 18 | 1775 | 101 |