Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRBP1
Entrez Gene ID
6238
Gene Name
ribosome binding protein 1
Gene Aliases
ES/130, ES130, RRp, hES
Chromosome Location
Chr.20: 17613678 - 17682283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004587.2 NP_004578.2 1 - 2 257 73
GenBank mRNA AF006751.1 - 1 - 2 50 73
BC009700.1 - 1 - 3 193 73

View Full Product Details