The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042576.1 | NP_001036041.1 | 6 - 7 | 1243 | 66 |
NM_004587.2 | NP_004578.2 | 5 - 6 | 1166 | 66 | |
GenBank mRNA | AB037819.2 | - | - | 2922 | 66 |
AB287347.2 | - | 2 - 3 | 2578 | 66 | |
AF006751.1 | - | 5 - 6 | 959 | 66 | |
AL833822.1 | - | 3 - 4 | 954 | 66 | |
BC020482.1 | - | 2 - 3 | 191 | 66 | |
BC080526.1 | - | 1 - 2 | 50 | 66 | |
BC128577.1 | - | 5 - 6 | 1352 | 66 | |
BC128578.1 | - | 4 - 5 | 386 | 66 |