Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCF2
Entrez Gene ID
4688
Gene Name
neutrophil cytosolic factor 2
Gene Aliases
NCF-2, NOXA2, P67-PHOX, P67PHOX
Chromosome Location
Chr.1: 183555562 - 183590921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000433.3 NP_000424.2 13 - 14 1559 113
NM_001127651.2 NP_001121123.1 14 - 15 1396 113
NM_001190789.1 NP_001177718.1 11 - 12 1316 113
NM_001190794.1 NP_001177723.1 12 - 13 1424 113
XM_005245207.1 XP_005245264.1 12 - 13 1380 113
XM_011509580.1 XP_011507882.1 14 - 15 1397 113
XM_011509581.1 XP_011507883.1 14 - 15 1384 113
GenBank mRNA AB209647.1 - 11 - 12 4829 113
AF527950.1 - 13 - 14 1306 113
AK296672.1 - 11 - 12 1172 113
AK298713.1 - 12 - 13 1333 113
AK312666.1 - 13 - 14 1469 113
BC001606.1 - 13 - 14 1536 113
BT007439.1 - 13 - 14 1284 113
KU178216.1 - 13 - 14 1284 113
M32011.1 - 14 - 15 1351 113

View Full Product Details