The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000433.3 | NP_000424.2 | 7 - 8 | 985 | 81 |
NM_001127651.2 | NP_001121123.1 | 8 - 9 | 822 | 81 | |
NM_001190789.1 | NP_001177718.1 | 5 - 6 | 742 | 81 | |
NM_001190794.1 | NP_001177723.1 | 6 - 7 | 850 | 81 | |
XM_005245207.1 | XP_005245264.1 | 6 - 7 | 806 | 81 | |
XM_011509580.1 | XP_011507882.1 | 8 - 9 | 823 | 81 | |
XM_011509581.1 | XP_011507883.1 | 8 - 9 | 810 | 81 | |
GenBank mRNA | AF527950.1 | - | 7 - 8 | 732 | 81 |
AK296672.1 | - | 5 - 6 | 598 | 81 | |
AK298713.1 | - | 6 - 7 | 759 | 81 | |
AK312666.1 | - | 7 - 8 | 895 | 81 | |
BC001606.1 | - | 7 - 8 | 962 | 81 | |
BT007439.1 | - | 7 - 8 | 710 | 81 | |
KU178216.1 | - | 7 - 8 | 710 | 81 | |
M32011.1 | - | 8 - 9 | 777 | 81 |