The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277813.1 | NP_001264742.1 | 1 - 2 | 345 | 289 |
NM_001277815.1 | NP_001264744.1 | 1 - 2 | 345 | 174 | |
NM_152758.5 | NP_689971.4 | 1 - 2 | 345 | 143 | |
NR_102434.1 | 1 - 2 | 345 | 143 | ||
GenBank mRNA | AK056219.1 | - | 1 - 2 | 255 | 143 |
AK127574.1 | - | 1 - 2 | 277 | 143 | |
AL832005.1 | - | 1 - 2 | 278 | 143 | |
AL832335.1 | - | 1 - 2 | 243 | 174 | |
BC052970.1 | - | 1 - 2 | 294 | 143 | |
BP223018.1 | - | 1 - 2 | 143 | ||
BX537996.2 | - | 1 - 2 | 282 | 143 | |
CN335163.1 | - | 1 - 2 | 332 | 143 | |
DC419051.1 | - | 1 - 2 | 277 | 289 |