Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PVALB
Entrez Gene ID
5816
Gene Name
parvalbumin
Gene Aliases
D22S749
Chromosome Location
Chr.22: 36800701 - 36819473 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC096112.3 - 1 - 2 46 80
BC096114.1 - 1 - 2 46 80
BC096115.1 - 1 - 2 46 80
X63070.1 - 1 - 2 46 80

View Full Product Details