The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098.2 | NP_001089.1 | 13 - 14 | 1633 | 71 |
XM_017028812.1 | XP_016884301.1 | 13 - 14 | 1947 | 71 | |
GenBank mRNA | AK293562.1 | - | 9 - 10 | 855 | 71 |
AK296260.1 | - | 14 - 15 | 1598 | 71 | |
AK297213.1 | - | 13 - 14 | 1508 | 71 | |
AK301321.1 | - | 8 - 9 | 774 | 71 | |
AK302691.1 | - | 14 - 15 | 1583 | 71 | |
AK314845.1 | - | 13 - 14 | 1641 | 71 | |
BC014092.2 | - | 13 - 14 | 1633 | 71 | |
BC026196.2 | - | 13 - 14 | 1636 | 71 | |
CR456365.1 | - | 13 - 14 | 1625 | 71 | |
CR536568.1 | - | 13 - 14 | 1611 | 71 | |
GQ472196.1 | - | 13 - 14 | 1636 | 71 |