Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFNG
Entrez Gene ID
4242
Gene Name
MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases
-
Chromosome Location
Chr.22: 37469063 - 37486440 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002405.3 NP_002396.2 2 - 3 569 69
GenBank mRNA BC094814.1 - 2 - 3 530 69
BM457814.1 - 2 - 3 524 69
CR456518.1 - 2 - 3 362 69
U94352.1 - 2 - 3 476 69

View Full Product Details