Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYB5D1
Entrez Gene ID
124637
Gene Name
cytochrome b5 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.17: 7857746 - 7864417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144607.4 NP_653208.2 3 - 4 840 81
GenBank mRNA AK289520.1 - 3 - 4 523 81
BC060779.1 - 3 - 4 592 81

View Full Product Details