Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPHN1
Entrez Gene ID
4983
Gene Name
oligophrenin 1
Gene Aliases
ARHGAP41, MRX60, OPN1
Chromosome Location
Chr.X: 68042344 - 68433841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002547.2 NP_002538.1 18 - 19 1814 100
XM_005262270.1 XP_005262327.1 18 - 19 1884 100
XM_006724653.2 XP_006724716.1 18 - 19 1884 100
XM_011530961.1 XP_011529263.1 18 - 19 1735 100
XM_017029555.1 XP_016885044.1 18 - 19 1884 100
GenBank mRNA AB102656.1 - 17 - 18 1520 100
AJ001189.1 - 18 - 19 2158 100
AK295769.1 - 18 - 19 1884 100
BC140763.1 - 18 - 19 1852 100

View Full Product Details