The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002547.2 | NP_002538.1 | 22 - 23 | 2618 | 101 |
XM_005262270.1 | XP_005262327.1 | 21 - 22 | 2364 | 101 | |
XM_006724653.2 | XP_006724716.1 | 22 - 23 | 2688 | 101 | |
XM_011530961.1 | XP_011529263.1 | 22 - 23 | 2539 | 101 | |
GenBank mRNA | AB102656.1 | - | 21 - 22 | 2324 | 101 |
AJ001189.1 | - | 22 - 23 | 2962 | 101 | |
AK295769.1 | - | 21 - 22 | 2364 | 101 | |
BC140763.1 | - | 22 - 23 | 2656 | 101 |