The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020242.2 | NP_064627.1 | 24 - 25 | 3090 | 69 |
XM_006713264.3 | XP_006713327.1 | 15 - 16 | 1948 | 69 | |
XM_011533964.2 | XP_011532266.1 | 23 - 24 | 2895 | 69 | |
XM_017006884.1 | XP_016862373.1 | 25 - 26 | 3162 | 69 | |
XM_017006885.1 | XP_016862374.1 | 24 - 25 | 3012 | 69 | |
XM_017006886.1 | XP_016862375.1 | 26 - 27 | 3210 | 69 | |
XM_017006887.1 | XP_016862376.1 | 25 - 26 | 3162 | 69 | |
XM_017006888.1 | XP_016862377.1 | 22 - 23 | 2829 | 69 | |
XM_017006889.1 | XP_016862378.1 | 25 - 26 | 3162 | 69 | |
XM_017006890.1 | XP_016862379.1 | 16 - 17 | 2017 | 69 | |
GenBank mRNA | AB035898.1 | - | 24 - 25 | 3014 | 69 |
AF308294.1 | - | 6 - 7 | 587 | 69 | |
AK027816.1 | - | 6 - 7 | 696 | 69 | |
AK307674.1 | - | 24 - 25 | 3009 | 69 | |
AL832908.1 | - | 15 - 16 | 1929 | 69 | |
BC117174.1 | - | 23 - 24 | 2943 | 69 | |
BC143752.1 | - | 23 - 24 | 2944 | 69 |