Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAGLB
Entrez Gene ID
221955
Gene Name
diacylglycerol lipase beta
Gene Aliases
DAGLBETA, KCCR13L
Chromosome Location
Chr.7: 6409116 - 6448012 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142936.1 NP_001136408.1 2 - 3 421 88
NM_139179.3 NP_631918.3 2 - 3 421 88
GenBank mRNA AF450090.1 - 2 - 3 363 88
AK074584.1 - 2 - 3 341 88
AK074744.1 - 2 - 3 361 88
AK075128.1 - 2 - 3 360 88
AK298955.1 - 2 - 3 344 88
BC027603.1 - 2 - 3 300 88
DC303993.1 - 2 - 3 407 88
DC304654.1 - 2 - 3 425 88

View Full Product Details