The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139179.3 | NP_631918.3 | 3 - 4 | 586 | 98 |
GenBank mRNA | AF450090.1 | - | 3 - 4 | 528 | 98 |
AK074584.1 | - | 3 - 4 | 506 | 98 | |
AK074744.1 | - | 3 - 4 | 526 | 98 | |
AK075128.1 | - | 3 - 4 | 525 | 98 | |
BC027603.1 | - | 3 - 4 | 465 | 98 |