The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002980.2 | NP_002971.2 | 11 - 12 | 1369 | 122 |
XM_005263730.4 | XP_005263787.1 | 8 - 9 | 1823 | 122 | |
XM_011511621.2 | XP_011509923.1 | 11 - 12 | 1384 | 122 | |
XM_017004670.1 | XP_016860159.1 | 11 - 12 | 1835 | 122 | |
XM_017004671.1 | XP_016860160.1 | 10 - 11 | 1276 | 122 | |
XM_017004672.1 | XP_016860161.1 | 11 - 12 | 1199 | 122 | |
XM_017004673.1 | XP_016860162.1 | 7 - 8 | 742 | 122 | |
GenBank mRNA | AK055551.1 | - | 8 - 9 | 1843 | 122 |
AK314638.1 | - | 11 - 12 | 1372 | 122 | |
AY462218.1 | - | 11 - 12 | 1143 | 122 | |
BC035757.1 | - | 11 - 12 | 1408 | 122 | |
U13989.1 | - | 11 - 12 | 1193 | 122 | |
U20178.1 | - | 11 - 12 | 122 | ||
U28281.1 | - | 11 - 12 | 1251 | 122 |