The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002980.2 | NP_002971.2 | 5 - 6 | 733 | 68 |
XM_011511621.2 | XP_011509923.1 | 5 - 6 | 733 | 68 | |
XM_017004670.1 | XP_016860159.1 | 5 - 6 | 1184 | 68 | |
XM_017004671.1 | XP_016860160.1 | 4 - 5 | 625 | 68 | |
XM_017004672.1 | XP_016860161.1 | 5 - 6 | 548 | 68 | |
GenBank mRNA | AK314638.1 | - | 5 - 6 | 736 | 68 |
AY462218.1 | - | 5 - 6 | 507 | 68 | |
BC035757.1 | - | 5 - 6 | 772 | 68 | |
U13989.1 | - | 5 - 6 | 557 | 68 | |
U20178.1 | - | 5 - 6 | 564 | 68 | |
U28281.1 | - | 5 - 6 | 615 | 68 |