The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052947.3 | NP_443179.3 | 9 - 10 | 6153 | 124 |
XM_011525801.2 | XP_011524103.1 | 9 - 10 | 6386 | 124 | |
XM_011525802.2 | XP_011524104.1 | 7 - 8 | 6773 | 124 | |
XM_017025535.1 | XP_016881024.1 | 9 - 10 | 6387 | 124 | |
XM_017025536.1 | XP_016881025.1 | 9 - 10 | 6387 | 124 | |
XM_017025538.1 | XP_016881027.1 | 6 - 7 | 5271 | 124 | |
XM_017025539.1 | XP_016881028.1 | 6 - 7 | 4019 | 124 | |
GenBank mRNA | AL832018.2 | - | 6 - 7 | 4485 | 124 |
AY044450.1 | - | 6 - 7 | 4220 | 124 | |
BX647446.1 | - | 5 - 6 | 3631 | 124 | |
BX647639.1 | - | 6 - 7 | 5019 | 124 | |
BX647796.1 | - | 9 - 10 | 6140 | 124 |