Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TDRKH
Entrez Gene ID
11022
Gene Name
tudor and KH domain containing
Gene Aliases
TDRD2
Chromosome Location
Chr.1: 151769354 - 151790576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083963.1 NP_001077432.1 8 - 9 1372 66
NM_001083964.1 NP_001077433.1 8 - 9 1262 66
NM_001083965.1 NP_001077434.1 8 - 9 1397 66
NM_006862.3 NP_006853.2 8 - 9 1397 66
XM_005244856.4 XP_005244913.1 8 - 9 1163 66
XM_017000122.1 XP_016855611.1 8 - 9 1439 66
XM_017000123.1 XP_016855612.1 8 - 9 1439 66
XM_017000124.1 XP_016855613.1 8 - 9 1338 66
XM_017000125.1 XP_016855614.1 8 - 9 1367 66
XM_017000126.1 XP_016855615.1 8 - 9 1338 66
XM_017000127.1 XP_016855616.1 8 - 9 1367 66
XM_017000128.1 XP_016855617.1 8 - 9 1292 66
XM_017000129.1 XP_016855618.1 8 - 9 1257 66
XM_017000130.1 XP_016855619.1 8 - 9 1231 66
GenBank mRNA AF119121.2 - 8 - 9 1385 66
AF227192.1 - 8 - 9 1385 66
AK056402.1 - 8 - 9 1320 66
AK225160.1 - 8 - 9 1251 66
AK226092.1 - 8 - 9 1310 66
AK295948.1 - 8 - 9 1168 66
AK303063.1 - 8 - 9 66
BC032690.1 - 8 - 9 1304 66

View Full Product Details