Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PNPLA8
Entrez Gene ID
50640
Gene Name
patatin like phospholipase domain containing 8
Gene Aliases
IPLA2-2, IPLA2G, MMLA, PNPLA-gamma, iPLA2gamma
Chromosome Location
Chr.7: 108470417 - 108528161 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256007.2 NP_001242936.1 3 - 4 1478 89
NM_001256008.2 NP_001242937.1 2 - 3 1432 89
NM_001256009.2 NP_001242938.1 2 - 3 1432 89
NM_001256010.2 NP_001242939.1 2 - 3 1305 89
NM_001256011.2 NP_001242940.1 3 - 4 1166 89
NM_015723.4 NP_056538.1 4 - 5 1590 89
XM_005250396.4 XP_005250453.1 3 - 4 2434 89
XM_011516274.2 XP_011514576.1 3 - 4 2307 89
XM_011516275.2 XP_011514577.1 3 - 4 2307 89
XM_017012261.1 XP_016867750.1 2 - 3 2261 89
GenBank mRNA AB041261.1 - 2 - 3 1258 89
AF263613.1 - 3 - 4 1284 89
AK024335.1 - 3 - 4 1166 89
AK291755.1 - 3 - 4 1295 89
AL834147.1 - 2 - 3 1185 89
BC032999.2 - 4 - 5 1466 89
BX647865.1 - 2 - 3 1151 89

View Full Product Details